missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC29A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565424
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: YARYYMRPVLA AHVFSGEEEL PQDSLSAPSV ASRFIDSHTP PLRPILKKT Highest antigen sequence identity to the following orthologs - mouse 64%, rat 62%.
SLC29A3 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A3 is a transmembrane glycoprotein that localizes to the lysosomal membrane and is a broad selectivity, low affinity nucleoside transporter. Mutations in the SLC29A3 gene have been associated with H syndrome, which is characterized by cutaneous hyperpigmentation and hypertrichosis, hepatosplenomegaly, heart anomalies, and hypogonadism. A related disorder, PHID (pigmented hypertrichosis with insulin-dependent diabetes mellitus), has also been associated with mutations at this locus.
Spécification
| SLC29A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC29A3 | |
| 4933435C21Rik; AW987637; Ent3; Equilibrative nucleoside transporter 3; HCLAP; hENT3; HJCD; mENT3; PHID; Slc29a3; solute carrier family 29 (equilibrative nucleoside transporter), member 3; solute carrier family 29 (nucleoside transporters), member 3; solute carrier family 29 member 3; UNQ717/PRO1380 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55315 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZD2 | |
| SLC29A3 | |
| Recombinant Human SLC29A3. Recombinant protein control fragment (Product #RP-95808). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu