missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC29A3 Polyclonal antibody specifically detects SLC29A3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | SLC29A3 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | equilibrative nucleoside transporter 3, ENT3, HCLAP, HJCD, PHID, solute carrier family 29 (nucleoside transporters), member 3 |
| Host Species | Rabbit |
| Immunogen | SLC29A3 antibody was raised against a 19 amino acid peptide near the amino terminus of human SLC29A3. The immunogen is located within the first 50 amino acids of SLC29A3. Amino Acid Squence: LEKLLDRPPPGLQRPEDRF |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?