missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC28A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101897
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC28A2.
The concentrative nucleoside transporter (CNT) family comprises three members: CNT1, CNT2 and CNT3. CNT2 participates in the absorption and disposition of endogenous nucleosides and mediates the first step of nucleotide biosynthesis. CNT2 levels are highly dependent on insulin (but not glucose) concentration, and the protein is under the control of the Adenosine 1 receptor. CNT family members are imperative in the response of cells to a variety of anticancer and antiviral nucleoside analogs, as the CNT proteins modulate their entry into target tissues. Increasing evidence also suggests that CNT2 may have a role in energy metabolism because activation of CNT2 relies on the opening of ATP-sensitive K+ channels.
Specifications
| SLC28A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC28A2 | |
| 2010208B10Rik; B430217P18; BB152493; CNT 2; CNT2; CNT-2; Concentrative nucleoside transporter 2; HCNT2; HsT17153; Na(+)/nucleoside cotransporter 2; purine-selective Na+ nucleoside cotransporter; rCNT2; Slc28a2; sodium/nucleoside cotransporter 2; sodium/purine nucleoside cotransporter; Sodium/purine nucleoside co-transporter; Sodium-coupled nucleoside transporter 2; solute carrier family 28 (concentrative nucleoside transporter), member 2; solute carrier family 28 (sodium-coupled nucleoside transporter), member 2; solute carrier family 28 member 2; solute carrier family 28, member 2; SPNT; SPNT1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 269346, 60423, 9153 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43868, O88627, Q62773 | |
| SLC28A2 | |
| A synthesized peptide derived from human SLC28A2(Accession O43868), corresponding to amino acid residues C372-N422. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction