missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC27A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581478
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse SLC27A3.
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
Specifications
| SLC27A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC27A3 | |
| Acsvl3; FATP3; FATP-3; fatty acid transport protein 3; long-chain fatty acid transport protein 3; mFLJ00207 protein; PSEC0067; Slc27a3; solute carrier family 27 (fatty acid transporter), member 3; solute carrier family 27 member 3; UNQ367/PRO703; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 26568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| O88561 | |
| SLC27A3 | |
| A synthetic peptide corresponding to the center region of the Mouse SLC27A3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction