missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC27A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566931
This item is not returnable.
View return policy
Description
Immunogen sequence: DTWERFVRRFG PLQVLETYGL TEGNVATINY TGQRGAVGRA SWLYKHIFPF SLIRYDVTTG EPIRDPQGHC MATSPGEPGL LVAPVSQQSP FLGYAGGPEL AQGKLLKDVF R Highest antigen sequence identity to the following orthologs - mouse 88%, rat 89%.
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
Specifications
| SLC27A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC27A3 | |
| Acsvl3; FATP3; FATP-3; fatty acid transport protein 3; long-chain fatty acid transport protein 3; mFLJ00207 protein; PSEC0067; Slc27a3; solute carrier family 27 (fatty acid transporter), member 3; solute carrier family 27 member 3; UNQ367/PRO703; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5K4L6 | |
| SLC27A3 | |
| Recombinant Human SLC27A3. Recombinant protein control fragment (Product #RP-107584). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido