Learn More
Invitrogen™ SLC27A3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA542490
Product Code. 15565502
  • £449.00 / 100µL
Description
Peptide sequence: PFSLIRYDVT TGEPIRDPQG HCMATSPGEP GLLVAPVSQQ SPFLGYAGGP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Sheep: 100%.
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC27A3
Polyclonal
Unconjugated
SLC27A3
Acsvl3; FATP3; FATP-3; fatty acid transport protein 3; long-chain fatty acid transport protein 3; mFLJ00207 protein; PSEC0067; Slc27a3; solute carrier family 27 (fatty acid transporter), member 3; solute carrier family 27 member 3; UNQ367/PRO703; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3
Rabbit
Affinity Chromatography
RUO
11000
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q5K4L6
SLC27A3
Synthetic peptide directed towards the middle region of human SLC27A3.