missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC27A2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588033

Product Code. 16596371

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TKFSASQFWD DCRKYNVTVI QYIGELLRYL CNSPQKPNDR DHKVRLALGN GLRGDVWRQF VKRFGDICIY EFYAATEGNI GFMNYARKVG AVGRVNYLQK KIITYDLIKY DVEKDEPVRD ENGYCVRVPK GEVGLLVCKI TQLTPFNGYA GAKAQTEKKK LRDVFKKGDL YFNSGDLLMV DHENFIYFHD RVGDTFRWKG ENVATTEVAD TVGLVDFVQE VNVYGVHVPD HEGRIGMASI KMKENHEFDG KKLFQHIADY LPSYARPRFL RIQDTIEITG TFKH Positive Samples: HepG2; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein, Peroxisome membrane.

The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC27A2
Polyclonal
Unconjugated
SLC27A2
ACSVL1; Arachidonate--CoA ligase; Facvl1; FATP2; FATP-2; Fatty acid transport protein 2; fatty acid transporter; fatty-acid-coenzyme A ligase, very long-chain 1; hFACVL1; HsT17226; Long-chain-fatty-acid--CoA ligase; Phytanate--CoA ligase; SLC27A2; solute carrier family 27 (fatty acid transporter) member 2; solute carrier family 27 (fatty acid transporter), member 2; solute carrier family 27 (fatty acid transporter), member 32; solute carrier family 27 member 2; THCA-CoA ligase; ve; very long-chain acyl-CoA synthetase; very long-chain acyl-Coenzyme A dehydrogenase synthase; very long-chain fatty-acid-coenzyme A ligase 1; very long-chain-fatty-acid-CoA ligase; very-long-chain acyl-CoA synthetase; Vlac; VLACS; Vlcs
Rabbit
Affinity Chromatography
RUO
11001, 26458, 65192
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.65 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
O14975, O35488, P97524
SLC27A2
Recombinant fusion protein containing a sequence corresponding to amino acids 290-573 of human FATP2 (NP_0036362).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado