missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98638CL1
This item is not returnable.
View return policy
Description
SLC26A6 Polyclonal antibody specifically detects SLC26A6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC26A6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| anion transporter 1, DKFZp586E1422, pendrin L1, pendrin-L1, Pendrin-like protein 1, solute carrier family 26 member 6, solute carrier family 26, member 6, sulfate anion transporter | |
| Produced in rabbits immunized with E. coli-derived Human SLC26A6 fragment. | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 65010 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction