missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93440-0.02ml
This item is not returnable.
View return policy
Description
SLC26A6 Polyclonal antibody specifically detects SLC26A6 in Human samples. It is validated for Western Blot
Specifications
| SLC26A6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| anion transporter 1, DKFZp586E1422, pendrin L1, pendrin-L1, Pendrin-like protein 1, solute carrier family 26 member 6, solute carrier family 26, member 6, sulfate anion transporter | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 509-758 of human SLC26A6 (NP_599025.2). QMPHYSVLGQVPDTDIYRDVAEYSEAKEVRGVKVFRSSATVYFANAEFYSDALKQRCGVDVDFLISQKKKLLKKQEQLKLKQLQKEEKLRKQAASPKGASVSINVNTSLEDMRSNNVEDCKMMVSSGDKMEDATANGQEDSKAPDGSTLKALGLPQPDFHSLILDLGALSFVDTVCLKSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRL | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 65010 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction