missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £251.00
Specifications
| Antigen | SLC26A2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651678
|
Novus Biologicals
NBP3-12557-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675167
|
Novus Biologicals
NBP3-12557-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC26A2 Polyclonal antibody specifically detects SLC26A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| SLC26A2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction | |
| PBS, pH7.0 | |
| 1836 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| D5S1708, Diastrophic dysplasia protein, diastrophic dysplasia sulfate transporter, DTDMSTP157, DTDSTMST153, EDM4, solute carrier family 26 (sulfate transporter), member 2, Solute carrier family 26 member 2, sulfate anion transporter 1, sulfate transporter | |
| Produced in rabbits immunized with E. coli-derived Human SLC26A2 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title