missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93651-0.1ml
This item is not returnable.
View return policy
Description
SLC26A2 Polyclonal antibody specifically detects SLC26A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SLC26A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| D5S1708, Diastrophic dysplasia protein, diastrophic dysplasia sulfate transporter, DTDMSTP157, DTDSTMST153, EDM4, solute carrier family 26 (sulfate transporter), member 2, Solute carrier family 26 member 2, sulfate anion transporter 1, sulfate transporter | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human SLC26A2 (NP_000103.2). MSSESKEQHNVSPRDSAEGNDSYPSGIHLELQRESSTDFKQFETNDQCRPYHRILIERQEKSDTNFKEFVIKKLQKNCQCSPAKAKNMILGFLPVLQWLPKYDLKKNILGD | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction | |
| 1836 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction