missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A6 Polyclonal Antibody

SLC25A6 Polyclonal Antibody
Brand: Thermo Scientific PA5106593
This item is not returnable.
View return policy
Description
SLC25A6 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| SLC25A6 | |
| Polyclonal | |
| Unconjugated | |
| Slc25a6 | |
| 4930433D19Rik; 4933433F13Rik; AAC3; adenine nucleotide translocator 2; adenine nucleotide translocator 3; adenine nucleotide translocator 3 (liver); ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocase 3, N-terminally processed; ADP/ATP translocator of liver; ANT; ANT 2; ANT 3; ANT3; ANT3Y; AW557176; CDABP0051; D6Bwg0781e; mitochondrial ADP/ATP carrier protein; mitochondrial S-adenosylmethionine transporter; mitochondrial solute carrier family 25 member 6; S-adenosylmethionine mitochondrial carrier protein; Samc; si:dkey-21o13.4; Slc25a26; slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; solute carrier family 25 member 26; Solute carrier family 25 member 6; solute carrier family 25, member A6; translocase; wu:fj78b08 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 293, 498885, 67582 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12236, Q5U680 | |
| Slc25a6 | |
| A synthesized peptide derived from human SLC25A6(Accession P12236), corresponding to amino acid residues A123-L173. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction