missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A6, Mouse anti-Human, Clone: 4B9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000293-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: DFARTRLAADVGKSGTEREFRGLGDCLVKITKSDGIRGLYQGFSVSVQGIIIYRAAYFGVYDTAKGMLPDPKNTHSpecifications
| SLC25A6 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6 | |
| AAC3/ANT3/ANT3Y/MGC17525 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4B9 | |
| In 1x PBS, pH 7.4 | |
| NM_001636 | |
| SLC25A6 | |
| SLC25A6 (NP_001627, 135 a.a. ∼ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 293 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction