missing translation for 'onlineSavingsMsg'
Learn More

SLC25A6 Antibody (4B9), Novus Biologicals™

Product Code. 18393838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393838 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393838 Supplier Novus Biologicals Supplier No. H00000293M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC25A6 Monoclonal antibody specifically detects SLC25A6 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4B9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001627
Gene Alias Adenine nucleotide translocator 3, ADP, ADP/ATP translocase 3, ADP/ATP translocator of liver, ANT 2, ANT 3, ANT3AAC3, ANT3Y, ATP carrier protein, ATP carrier protein 3, ATP carrier protein, isoform T2, ATP carrier protein, liver, MGC17525, solute carrier family 25 (mitochondrial carrier, adenine nucleotidetranslocator), member 6, Solute carrier family 25 member 6
Host Species Mouse
Immunogen SLC25A6 (NP_001627, 135 a.a. ~ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DFARTRLAADVGKSGTEREFRGLGDCLVKITKSDGIRGLYQGFSVSVQGIIIYRAAYFGVYDTAKGMLPDPKNTH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.