missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A53 Polyclonal Antibody
SLC25A53 Polyclonal Antibody
Brand: Thermo Scientific PA557095
This item is not returnable.
View return policy
Description
SLC25A53 Polyclonal Antibody for IHC
Specifications
| SLC25A53 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A53 | |
| 2310046F18Rik; 2810402A17Rik; AI448995; AI841128; Mcart1l; MCART6; mitochondrial carrier triple repeat 1-like; mitochondrial carrier triple repeat 6; mitochondrial carrier triple repeat protein 6; Slc25a53; Solute carrier family 25 member 53; solute carrier family 25, member 53 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 401612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5H9E4 | |
| SLC25A53 | |
| Recombinant protein corresponding to Human SLC25A53. Recombinant protein control fragment (Product #RP-95466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction