missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A47 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26292-1-AP-20UL
This item is not returnable.
View return policy
Description
The SLC25A47 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC25A47. This antibody recognizes human, mouse antigen. The SLC25A47 antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.Specifications
| SLC25A47 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 283600 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 300 μg/mL | |
| HDMCP; HMFN1655 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Polyclonal Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction