missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A46 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20392
This item is not returnable.
View return policy
Description
SLC25A46 Polyclonal specifically detects SLC25A46 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLC25A46 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| solute carrier family 25 member 46, solute carrier family 25, member 46, TB1 | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q96AG3 | |
| SLC25A46 | |
| Recombinant protein encompassing a sequence within the center region of human SLC25A46. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 91137 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction