missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557189
This item is not returnable.
View return policy
Description
Immunogen sequence: ETVKRKMQAQ SPYLPHSGGV DVHFSGAVDC FRQIVKAQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Specifications
| SLC25A43 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A43 | |
| Gm8; mitochondrial solute carrier protein; RGD1559928; Slc25a43; Solute carrier family 25 member 43; solute carrier family 25, member 43 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 203427 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUT9 | |
| SLC25A43 | |
| Recombinant protein corresponding to Human SLC25A43. Recombinant protein control fragment (Product #RP-95459). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction