missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589088
This item is not returnable.
View return policy
Description
Positive Samples: 293T; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein Immunogen sequence: MIQNSRPSLL QPQDVGDTVE TLMLHPVIKA FLCGSISGTC STLLFQPLDL LKTRLQTLQP SDHGSRRVGM LAVLLKVVRT ESLLGLWKGM SPSIVRCVPG.
SLC25A38 is a members of the solute carrier family 25 (SLC25) which is known to transport molecules over the mitochondrial membrane.
Specifications
| SLC25A38 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A38 | |
| appoptosin; AV019094; BC010801; HAMAP-Rule:MF_03064}; hypothetical protein FLJ20551; mitochondrial glycine transporter; Mitochondrial glycine transporter GlyC; RGD1311914; SIDBA2; Slc25a38; solute carrier family 25 member 38; solute carrier family 25 member 38 {ECO:0000255; solute carrier family 25, member 38 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 208638, 54977 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.72 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q91XD8, Q96DW6 | |
| SLC25A38 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human SLC25A38 (NP_0603452). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction