missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569293
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 91%.
SLC25A34 belongs to the SLC25 family of mitochondrial carrier proteins.
Specifications
| SLC25A35 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A35 | |
| 1810012H11Rik; si:ch211-268m12.5; similar to 1810012H11Rik; slc25a35; Solute carrier family 25 member 35; solute carrier family 25, member 35 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 399512 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3KQZ1 | |
| SLC25A35 | |
| synthetic peptide directed towards the middle region of human SLC25A35. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction