missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17557-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC25A34 belongs to the SLC25 family of mitochondrial carrier proteins.Specifications
| SLC25A34 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A34 | |
| RP11 169K16.2, SLC25A34 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 284723, 298606, 384071 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2ADF7, Q5XIF9, Q6PIV7 | |
| SLC25A34 | |
| SLC25A34 Fusion Protein Ag11719 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction