missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582520
This item is not returnable.
View return policy
Description
Immunogen sequence: FARTRLGVDI GKGPEERQFK GLGDCIMKIA KSDGIAGLYQ GFGVSVQGII VYRA Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.
Mitochondrial ADP/ATP carriers, such as SLC25A31, are nuclear-coded mitochondrial proteins that catalyze the exchange of ATP generated in mitochondria by ATP synthase (see MIM 108729) against ADP produced in cytosol by most energy-consuming reactions.
Specifications
| SLC25A31 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A31 | |
| 1700034J06Rik; Aac4; adenine nucleotide translocase 4; adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 4; ADP/ATP translocase 4; ANT 4; Ant4; LOC541168 protein; Sfec; SFEC35kDa; SLC25A31; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 member 31; Sperm flagellar energy carrier protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83447 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0C2 | |
| SLC25A31 | |
| Recombinant protein corresponding to Human SLC25A31. Recombinant protein control fragment (Product #RP-90992). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction