missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SLC25A31 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC25A31 Polyclonal specifically detects SLC25A31 in Human samples. It is validated for Western Blot.Specifications
| SLC25A31 | |
| Polyclonal | |
| Rabbit | |
| Q9H0C2 | |
| 83447 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AAC4DKFZp434N1235, adenine nucleotide translocase 4, Adenine nucleotide translocator 4, ADP, ADP/ATP translocase 4, ANT 4, ANT4DKFZP434N1235, ATP carrier protein 4, SFEC, SFEC35kDa, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 31, Solute carrier family 25 member 31, Sperm flagellar energy carrier protein | |
| SLC25A31 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title