missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113047
This item is not returnable.
View return policy
Description
SLC25A25 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
Specifications
| SLC25A25 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A25 | |
| 1110030N17Rik; APC3; ATP-Mg/Pi carrier; calcium-binding mitochondrial carrier protein SCaMC-2; Kiaa1896; LOW QUALITY PROTEIN: calcium-binding mitochondrial carrier protein SCaMC-2; MCSC; MCSC3; Mitochondrial ATP-Mg/Pi carrier protein; mitochondrial ATP-Mg/Pi carrier protein 3; Mitochondrial Ca(2+)-dependent solute carrier protein 3; mitochondrial Ca2+-dependent solute carrier; mKIAA1896; Pcscl; peroxisomal Ca(2+)-dependent solute carrier-like protein; peroxisomal Ca-dependent solute carrier-like protein; Scamc2; SCAMC-2; short calcium-binding mitochondrial carrier 2; SLC25A25; small calcium-binding mitochondrial carrier 2; small calcium-binding mitochondrial carrier protein 2; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25; solute carrier family 25 member 25; Unknown (protein for MGC:127973); UNQ549/PRO1106 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 114789, 227731 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A2ASZ8, Q6KCM7 | |
| SLC25A25 | |
| Recombinant Human Calcium-binding mitochondrial carrier protein SCaMC-2 protein (1-189AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction