missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC25A25 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582724

Product Code. 16339684

  • £487.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LRLVFKSLDK KNDGRIDAQE IMQSLRDLGV KISEQQAEKI LKSMDKNGTM TIDWNEWRDY HLLHPVENIP EIILYWKHST IFDVGENLT Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC25A25
Polyclonal
Unconjugated
SLC25A25
1110030N17Rik; APC3; ATP-Mg/Pi carrier; calcium-binding mitochondrial carrier protein SCaMC-2; Kiaa1896; LOW QUALITY PROTEIN: calcium-binding mitochondrial carrier protein SCaMC-2; MCSC; MCSC3; Mitochondrial ATP-Mg/Pi carrier protein; mitochondrial ATP-Mg/Pi carrier protein 3; Mitochondrial Ca(2+)-dependent solute carrier protein 3; mitochondrial Ca2+-dependent solute carrier; mKIAA1896; Pcscl; peroxisomal Ca(2+)-dependent solute carrier-like protein; peroxisomal Ca-dependent solute carrier-like protein; Scamc2; SCAMC-2; short calcium-binding mitochondrial carrier 2; SLC25A25; small calcium-binding mitochondrial carrier 2; small calcium-binding mitochondrial carrier protein 2; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25; solute carrier family 25 member 25; Unknown (protein for MGC:127973); UNQ549/PRO1106
Rabbit
Antigen affinity chromatography
RUO
114789, 227731, 246771
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A2ASZ8, Q6KCM7, Q8K3P6
SLC25A25
Recombinant protein corresponding to Human SLC25A25. Recombinant protein control fragment (Product #RP-93011).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.