missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14669-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC25A24 is a calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. It may act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.Specifications
| SLC25A24 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A24 | |
| APC1, MCSC1, SCAMC 1, SCAMC1, SLC25A24 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229731, 29957, 310791 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6NUK1, Q8BMD8 | |
| SLC25A24 | |
| SLC25A24 Fusion Protein Ag6334 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction