missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35221-20ul
This item is not returnable.
View return policy
Description
SLC25A24 Polyclonal antibody specifically detects SLC25A24 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| SLC25A24 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| APC1calcium-binding mitochondrial carrier protein SCaMC-1, calcium-binding transporter, DKFZp586G0123, MCSC1, Mitochondrial ATP-Mg/Pi carrier protein 1, mitochondrial ATP-Mg/Pi transporter, Mitochondrial Ca(2+)-dependent solute carrier protein 1, SCAMC1, SCAMC-1, short calcium-binding mitochondrial carrier 1, Small calcium-binding mitochondrial carrier protein 1, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24, Solute carrier family 25 member 24 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SLC25A24 (NP_037518.3).,, Sequence:, MLRWLRDFVLPTAACQDAEQPTRYETLFQALDRNGDGVVDIGELQEGLRNLGIPLGQDAEEKIFTTGDVNKDGKLDFEEFMKYLKDHEKKMKLAFKSLDKNNDGKIEASEIVQSLQTLGLTISEQQAELILQSIDVDGTMTVDWNEWRDYFLFNPVTDIEEIIRFWKHSTGIDIGDSLTIPDEFTEDEKKSGQWWRQLLA | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29957 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction