missing translation for 'onlineSavingsMsg'
Learn More

SLC25A24 Antibody (2C4), Novus Biologicals™

Product Code. 18386669 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386669 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386669 Supplier Novus Biologicals Supplier No. H00029957M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC25A24 Monoclonal antibody specifically detects SLC25A24 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A24
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_037518
Gene Alias APC1calcium-binding mitochondrial carrier protein SCaMC-1, calcium-binding transporter, DKFZp586G0123, MCSC1, Mitochondrial ATP-Mg/Pi carrier protein 1, mitochondrial ATP-Mg/Pi transporter, Mitochondrial Ca(2+)-dependent solute carrier protein 1, SCAMC1, SCAMC-1, short calcium-binding mitochondrial carrier 1, Small calcium-binding mitochondrial carrier protein 1, solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 24, Solute carrier family 25 member 24
Host Species Mouse
Immunogen SLC25A24 (NP_037518.2, 1 a.a. ~ 66 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLRWLRDFVLPTAACQDAEQPTRYETLFQALDRNGDGVVDIGELQEGLRNLGIPLGQDAEEKIFTT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29957
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.