missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564930
This item is not returnable.
View return policy
Description
Immunogen sequence: ELDSNKDGRVD VHELRQGLAR LGGGNPDPGA QQGISSEGDA DPDGGLDLEE FSRYLQEREQ RLLLMFHSLD RNQDGHIDVS EIQQSFRALG ISISLEQAEK ILHSMDRDGT MTIDWQEWRD HFLLHSL Highest antigen sequence identity to the following orthologs - mouse 93%, rat 90%.
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane (PubMed:15123600). May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria (PubMed:15123600). Acts as a regulator of mitochondrial calcium uptake via interaction with MCU and MICU1 (PubMed:24430870). [UniProt].
Specifications
| SLC25A23 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A23 | |
| 2310067G05Rik; APC2; calcium-binding mitochondrial carrier protein SCaMC-3; MCSC2; mitochondrial ATP-Mg/Pi carrier protein 2; Mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; phosphate carrier), member 23; SCAMC3; SCaMC-3; short calcium-binding mitochondrial carrier 3; Slc25a23; sma; Small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; solute carrier family 25 member 23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79085 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BV35 | |
| SLC25A23 | |
| Recombinant Human SLC25A23. Recombinant protein control fragment (Product #RP-105717). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction