Learn More
SLC25A22 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-25402-100UL
Description
The SLC25A22 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC25A22. This antibody recognizes human, mouse, rat antigen. The SLC25A22 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SLC25A22 | |
| 1000 μg/mL | |
| FLJ13044, GC 1, GC1, Glutamate/H(+) symporter 1, SLC25A22 | |
| 100 μL | |
| 79751 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |