missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553207
This item is not returnable.
View return policy
Description
Immunogen sequence: RSEGYFGMYR GAAVNLTLVT PEKAIKLAAN DFFRHQLSKD GQKLTLLKEM LAGCGAGTCQ VIVTTPMEML KIQLQDAGRI AAQRKILAAQ GQLSAQGGAQ PSVE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 94%.
The SLC25 gene family encodes mitochondrial carriers that transport a variety of metabolites across the inner mitochondrial membrane (Palmieri, 2004 ). SLC25A22, also known as GC1, is 1 of the 2 mitochondrial glutamate/H+ symporters, the other being SLC25A18.
Specifications
| SLC25A22 | |
| Polyclonal | |
| Unconjugated | |
| Slc25a22 | |
| 1300006L01Rik; AI060884; EIEE3; FLJ13044; GC1; GC-1; Glutamate/H(+) symporter 1; mitochondrial glutamate carrier 1; NET44; RGD1307826; Slc25a22; solute carrier family 25 (mitochondrial carrier, glutamate), member 22; solute carrier family 25 (mitochondrial carrier: glutamate), member 22; solute carrier family 25 member 22 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79751 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H936 | |
| Slc25a22 | |
| Recombinant protein corresponding to Human SLC25A22. Recombinant protein control fragment (Product #RP-90492). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction