missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SLC25A21 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC25A21 Polyclonal specifically detects SLC25A21 in Human samples. It is validated for Western Blot.Specifications
| SLC25A21 | |
| Polyclonal | |
| Rabbit | |
| Q9BQT8 | |
| 89874 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| MGC126570, ODC1, ODCmitochondrial 2-oxodicarboxylate carrier, oxodicarboxylate carrier, solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21, Solute carrier family 25 member 21 | |
| SLC25A21 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title