missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A20 Recombinant Rabbit Monoclonal Antibody (3J1L5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555555
This item is not returnable.
View return policy
Description
Immunogen Sequence: MADQPKPISP LKNLLAGGFG GVCLVFVGHP LDTVKVRLQT QPPSLPGQPP MYSGTFDCFR KTLFREGITG LYRGMAAPII GVTPMFAVCF FGFGLGKKLQ.
SLC25A20 is one of several closely related mitochondrial-membrane carrier proteins that shuttle substrates between cytosol and the intramitochondrial matrix space. It mediates the transport of acylcarnitines into mitochondrial matrix for their oxidation by the mitochondrial fatty acid-oxidation pathway. Mutations in this gene are associated with carnitine-acylcarnitine translocase deficiency, which can cause a variety of pathological conditions such as hypoglycemia, cardiac arrest, hepatomegaly, hepatic dysfunction and muscle weakness, and is usually lethal in new born and infants. This gene product is one of several closely related mitochondrial-membrane carrier proteins that shuttle substrates between cytosol and the intramitochondrial matrix space. This protein mediates the transport of acylcarnitines into mitochondrial matrix for their oxidation by the mitochondrial fatty acid-oxidation pathway. Mutations in this gene are associated with carnitine-acylcarnitine translocase deficiency, which can cause a variety of pathological conditions such as hypoglycemia, cardiac arrest, hepatomegaly, hepatic dysfunction and muscle weakness, and is usually lethal in new born and infants.
Specifications
| SLC25A20 | |
| Recombinant Monoclonal | |
| 1.02 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P97521, Q9Z2Z6 | |
| SLC25A20 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC25A20. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3J1L5 | |
| Unconjugated | |
| SLC25A20 | |
| 1110007P09Rik; C78826; CAC; CACT; carnitine/acylcarnitine carrier protein; carnitine/acylcarnitine translocase; mCAC; mitochondrial carnitine/acylcarnitine carrier protein; mitochondrial carnitine-acylcarnitine translocase; slc25a20; solute carrier family 25 (carnitine/acylcarnitine translocase), member 20; solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20; solute carrier family 25 member 20; Unknown (protein for MGC:128410); zgc:77760 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 117035, 57279 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction