missing translation for 'onlineSavingsMsg'
Learn More

SLC25A20 Antibody (M2), Novus Biologicals™

Product Code. 18383468 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383468 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383468 Supplier Novus Biologicals Supplier No. H00000788M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC25A20 Monoclonal antibody specifically detects SLC25A20 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A20
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone M2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01689
Gene Alias CACCarnitine/acylcarnitine translocase, CACTSolute carrier family 25 member 20, mitochondrial carnitine/acylcarnitine carrier protein, solute carrier family 25 (carnitine/acylcarnitine translocase), member 20
Host Species Mouse
Immunogen SLC25A20 (AAH01689, 1 a.a. ~ 301 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MADQPKPISPLKNLLAGGFGGVCLVFAGHPLDTVKVRLQTQPPSLPGQPPMYSGTFDCFRKTLFREGITGLYRGMAAPIIGVTPMFAVCFFGFGLGKKLQQKHPEDVLSYPQLFAAGMLSGVFTTGIMTPGERIKCLLQIQASSGESKYTGTLDCAKKLYQEFGIRGIYKGTVLTLMRDVPASGMYFMTYEWLKNIFTPEGKRVSELSAPRILVAGGIAGIFNWAVAIPPDVLKSRFQTAPPGKYPNGFRDVLRELIRDEGVTSLYKGFNAVMIRAFPANAACFLGFEVAMKFLNWATPNL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 788
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.