missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584124
This item is not returnable.
View return policy
Description
Immunogen sequence: TARLRLQVDE KRKSKTTHMV LLEIIKEEGL LAPYR Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.
This protein encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response.
Specifications
| SLC25A17 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A17 | |
| 34 kDa peroxisomal membrane protein; 34kDa; 47kDa; peroxisomal integral membrane protein 47; peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; Pmp34; Pmp35; Pmp47; SLC25A17; solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; solute carrier family 25 member 17; solute carrier family 25, member 17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10478 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43808 | |
| SLC25A17 | |
| Recombinant protein corresponding to Human SLC25A17. Recombinant protein control fragment (Product #RP-91536). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction