missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94298-0.1ml
This item is not returnable.
View return policy
Description
SLC25A17 Polyclonal antibody specifically detects SLC25A17 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SLC25A17 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| 34 kDa peroxisomal membrane protein, peroxisomal membrane protein (34kD), peroxisomal membrane protein PMP34, PMP34, Solute Carrier Family 25 (Mitochondrial Carrier; Peroxisomal Membrane Protein, 34kD), Member 17, Solute Carrier Family 25 Member 17 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SLC25A17 (NP_006349.1). IDAFHQIIRDEGISALWNGTFPSLLLVFNPAIQFMFYEGLKRQLLKKRMKLSSLDVFIIGAVAKAIATTVTYPLQTVQSILRFGRHRLNPENRTLGSLRNI | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10478 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction