missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A13, Mouse anti-Human, Clone: 4F4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010165-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: AAAKVALTKRADPAELRTIFLKYASIEKNGEFFMSPNDFVTRYLNIFGESQPNPKTVELLSGVVDQTKDGLISFQEFVASpecifications
| SLC25A13 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 25, member 13 (citrin) | |
| ARALAR2/CITRIN/CTLN2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, KnockDown, Western Blot | |
| 4F4 | |
| In 1x PBS, pH 7.4 | |
| NM_014251 | |
| SLC25A13 | |
| SLC25A13 (NP_055066, 2 a.a. ∼ 80 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10165 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction