missing translation for 'onlineSavingsMsg'
Learn More

SLC25A13 Antibody (4F4), Novus Biologicals™

Product Code. 18332929 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332929 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332929 Supplier Novus Biologicals Supplier No. H00010165M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC25A13 Monoclonal antibody specifically detects SLC25A13 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A13
Applications Western Blot, ELISA, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 4F4
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055066
Gene Alias ARALAR2CTLN2, calcium-binding mitochondrial carrier protein Aralar2, Citrin, Mitochondrial aspartate glutamate carrier 2, Solute carrier family 25 member 13, solute carrier family 25, member 13 (citrin)
Host Species Mouse
Immunogen SLC25A13 (NP_055066, 2 a.a. ~ 80 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AAAKVALTKRADPAELRTIFLKYASIEKNGEFFMSPNDFVTRYLNIFGESQPNPKTVELLSGVVDQTKDGLISFQEFVA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10165
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.