missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC25A12 Recombinant Rabbit Monoclonal Antibody (24GB2340)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen SLC25A12
Clone 24GB2340
Concentration 0.8 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274834
GREENER_CHOICE
View Documents
Invitrogen™
MA557714
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275317
View Documents
Invitrogen™
MA557715
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC25A12 Recombinant Monoclonal Antibody for Western Blot, Flow

This gene encodes a calcium-binding mitochondrial carrier protein. The encoded protein localizes to the mitochondria and is involved in the exchange of aspartate for glutamate across the inner mitochondrial membrane. Polymorphisms in the gene encoding this protein may be associated with risk of autism.
Specifications

Specifications

SLC25A12
0.8 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75746, Q8BH59
362145, 78830, 8604
A synthesized peptide derived from human SLC25A12 (aa 601-650).
Antibody
24GB2340
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
SLC25A12
2610002D09Rik; AGC1; AI839531; araceli hiperlarga; ARALAR; Aralar1; B230107K20Rik; BB129864; calcium binding mitochondrial carrier superfamily member Aralar1; calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; mitochondrial aspartate glutamate carrier 1; RGD1561141; SLC25A12; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; solute carrier family 25 member 12; solute carrier family 25, member 12
SLC25A12
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.