missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12253-1-AP-20UL
This item is not returnable.
View return policy
Description
The oxoglutarate/malate carrier transports 2-oxoglutarate across the inner membranes of mitochondria in an electroneutral exchange for malate or other dicarboxylic acids (summary by Iacobazzi et al., 1992 ).Specifications
| SLC25A11 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A11 | |
| OGC, OGCP, SLC20A4, SLC25A11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8402 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q02978 | |
| SLC25A11 | |
| SLC25A11 Fusion Protein Ag2903 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction