missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SLC25A11 Antibody (3G4), Novus Biologicals™

Product Code. 18347128 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347128 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347128 Supplier Novus Biologicals™ Supplier No. H00008402M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC25A11 Monoclonal antibody specifically detects SLC25A11 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A11
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 3G4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06508
Gene Alias OGCmitochondrial 2-oxoglutarate/malate carrier protein, OGCP, SLC20A4solute carrier family 20 (oxoglutarate carrier), member 4, solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member11, Solute carrier family 25 member 11
Host Species Mouse
Immunogen SLC25A11 (AAH06508, 1 a.a. ∽ 314 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAATASAGAGGIDGKPRTSPKSVKFLFGGLAGMGATVFVQPLDLVKNRMQLSGEGAKTREYKTSFHALTSILKAEGLRGIYTGLSAGLLRQATYTTTRLGIYTVLFERLTGADGTPPGFLLKAVIGMTAGATGAFVGTPAEVALIRMTADGRLPADQRRGYKNVFNALIRITREEGVLTLWRGCIPTMARAVVVNAAQLASYSQSKQFLLDSGYFSDNILCHFCASMISGLVTTAASMPVDIAKTRIQNMRMIDGKPEYKNGLDVLFKVVRYEGFFSLWKGFTPYYARLGPHTVLTFIFLEQMNKAYKRLFLSG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8402
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.