missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15235-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody specifically recognized the endogenous SLC25A1; no protein was detected in cells from subjects containing two alleles with truncating mutations with this antibody.
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.Specifications
| SLC25A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A1 | |
| CIC, Citrate transport protein, CTP, SLC20A3, SLC25A1, Tricarboxylate carrier protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13358, 29743, 6576 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32089, P53007, Q8JZU2 | |
| SLC25A1 | |
| SLC25A1 Fusion Protein Ag7352 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction