missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525403
This item is not returnable.
View return policy
Description
SLC25A1 Polyclonal Antibody for Western Blot, IHC (P), Flow
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
Specifications
| SLC25A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A1 | |
| 1300019P08Rik; 2610100G11Rik; AI194714; Cic; Citrate transport protein; citrate transporter) member 1; clathrin heavy chain 2; clathrin heavy chain 2; uncharacterized protein LOC100053137; clathrin, heavy chain-like 1; CLTCL1; Ctp; D2L2AD; Dgsj; DiGeorge syndrome gene j; LOC100053137; mitochondrial tricarboxylate carrier; SEA; Slc20a3; SLC25A1; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; solute carrier family 25 member 1; Tricarboxylate carrier protein; tricarboxylate transport protein, mitochondrial; txtp | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6576 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P53007 | |
| SLC25A1 | |
| KLH conjugated synthetic peptide between 142-170 amino acids from the central region of human TXTP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction