missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | SLC25A1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687791
|
Novus Biologicals
NBP2-93363-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679870
|
Novus Biologicals
NBP2-93363-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC25A1 Polyclonal antibody specifically detects SLC25A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SLC25A1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 6576 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Citrate transport protein, CTPTricarboxylate carrier protein, SEA, SLC20A3member 3, solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1, Solute carrier family 25 member 1, tricarboxylate transport protein, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-311 of human SLC25A1 (NP_005975.1). MPAPRAPRALAAAAPASGKAKLTHPGKAILAGGLAGGIEICITFPTEYVKTQLQLDERSHPPRYRGIGDCVRQTVRSHGVLGLYRGLSSLLYGSIPKAAVRFGMFEFLSNHMRDAQGRLDSTRGLLCGLGAGVAEAVVVVCPMETIKVKFIHDQTSPNPKYRGFFHGVREIVREQGLKGTYQGLTATVLKQGSNQAIRFFVMTSLRNWYRGDNPNKPMNPLITGVFGAIAGAASVFGNTPLDVIKTRMQGLEAHKYRNTWDCGLQILKKEGLKAFYKGTVPRLGRVCLDVAIVFVIYDEVVKLLNKVWKTD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title