missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC24A5 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518521
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine and human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
The SLC24A5 gene is a member of the potassium-dependent sodium/calcium exchanger family and encodes an intracellular membrane protein with 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments. Sequence variation in this gene has been associated with differences in skin pigmentation.
Specifications
| SLC24A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC24A5 | |
| F630045L20Rik; ion transporter JSX; JSX; K-independent Na+/Ca2+ exchanger JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; Na-Ca exchanger 5; NCKX5; NCX5; OCA6; oculocutaneous albinism 6 (autosomal recessive); SHEP4; Slc24a5; sodium/potassion/calcium exchanger 5; sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; solute carrier family 24 member 5; solute carrier family 24, member 5 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 283652, 311387, 317750 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q71RS6, Q8C261 | |
| SLC24A5 | |
| Synthetic peptide sequence (TTPDCRKKFWKN) corresponding to the internal amino acids of SLC24A5 (aa 321-332). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction