missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC24A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18992-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the potassium-dependent sodium/calcium exchanger protein family. Alternative splicing results in multiple transcript variants.Specifications
| SLC24A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC24A4 | |
| NCKX4, SHEP6, SLC24A2, SLC24A4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 123041, 238384 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CGQ8, Q8NFF2 | |
| Slc24a4 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction