missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC24A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539469
This item is not returnable.
View return policy
Description
Plasma membrane sodium/calcium exchangers are an important component of intracellular calcium homeostasis and electrical conduction. Potassium-dependent sodium/calcium exchangers such as SLC24A3 are believed to transport 1 intracellular calcium and 1 potassium ion in exchange for 4 extracellular sodium ions (Kraev et al., 2001 [PubMed 11294880]).[supplied by OMIM, Mar 2008].
Specifications
| SLC24A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC24A3 | |
| NCKX3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57419, 85267, 94249 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HC58, Q99PD7, Q9EPQ0 | |
| SLC24A3 | |
| A synthetic peptide derived from the internal region of human SLC24A3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction