missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC23A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113474
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SLC23A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Sodium/ascorbate cotransporter. Mediates electrogenic uptake of vitamin C, with a stoichiometry of 2 Na(+) for each ascorbate.
Spécification
| SLC23A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC23A1 | |
| D18Ucla2; hSVCT1; Na(+)/L-ascorbic acid transporter 1; SLC23A1; Slc23a2; Sodium-dependent vitamin C transporter 1; sodium-dependent vitamin C transporter-1; solute carrier family 23 (ascorbic acid transporter), member 1; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; solute carrier family 23 member 1; SVCT1; yolk sac permease-like molecule 3; YSPL3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 20522, 9963 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UHI7, Q9Z2J0 | |
| SLC23A1 | |
| Recombinant Human Solute carrier family 23 member 1 protein (173-259AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu