missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542232
This item is not returnable.
View return policy
Description
Peptide sequence: LSLPKLTYGG IALLAAGTAL LLPETRQAQL PETIQDVERK SAPTSLQEEE Sequence homology: Cow: 93%; Dog: 91%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
SLC22A7 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described. The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| SLC22A7 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A7 | |
| hOAT2; liver-specific transporter; MGC24091; MGC45202; mOAT2; NLT; novel liver transporter; oat2; organic anion transporter 2; rbOAT2; renal organic anion transporter 2; Slc22a7; solute carrier family 22 (organic anion transporter), member 7; solute carrier family 22 member 7; solute carrier family 22 member 7; LOW QUALITY PROTEIN: solute carrier family 22 member 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10864 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y694 | |
| SLC22A7 | |
| Synthetic peptide directed towards the C-terminal of human SLC22A7 (aa 491-540). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction