missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69696
This item is not returnable.
View return policy
Description
SLC22A7 Polyclonal specifically detects SLC22A7 in Human samples. It is validated for Western Blot.
Specifications
| SLC22A7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| hOAT2, liver-specific transporter, NLTMGC45202, Novel liver transporter, OAT2MGC24091, Organic anion transporter 2, solute carrier family 22 (organic anion transporter), member 7, solute carrier family 22 member 7 | |
| Rabbit | |
| 60 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y694-2 | |
| SLC22A7 | |
| Synthetic peptides corresponding to SLC22A7(solute carrier family 22 (organic anion transporter), member 7) The peptide sequence was selected from the N terminal of SLC22A7. Peptide sequence EPDGTLSSCLRFAYPQALPNTTLGEERQSRGELEDEPATVPCSQGWEYDH The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 10864 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction